Assay Details
Target Gene Details
Entrez Gene ID: | 1120 |
Gene Name: | choline kinase beta |
Gene Aliases: |
CHETK, CHKL, CK, CKB, CKEKB, EK, EKB, MDCMC |
Location: |
Chr.22:50578958-50582999 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHKB | NM_005198.4 | 2 | 523 | NP_005189.2 |
| AB029886.1 | 2 | 373 | BAA82512.1 | |
| AK097775.1 | 3 | 305 | ||
| AK314324.1 | 2 | 357 | ||
| AL096780.1 | 2 | 448 | CAB46629.1 | |
| AL096781.1 | 2 | 305 | CAB46630.1 | |
| BC082263.1 | 2 | 360 | AAH82263.1 | |
| BC101488.1 | 2 | 368 | ||
| BC113521.1 | 2 | 368 | ||
| CR456419.1 | 2 | 434 | CAG30305.1 | |
| DB106393.1 | 2 | 524 |
Target Gene Details
Entrez Gene ID: | 386593 |
Gene Name: | CHKB-CPT1B readthrough (NMD candidate) |
Gene Aliases: |
CHKL-CPT1B |
Location: |
Chr.22:50568861-50582999 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHKB-CPT1B | NR_027928.2 | 2 | 523 | |
| AB051457.1 | 1 | 689 | BAB33340.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059234 | Chr.22:50560544 - 50617472 on Build GRCh38 | Gain |
|
| nsv1057656 | Chr.22:50478783 - 50595520 on Build GRCh38 | Gain |
|
| nsv829340 | Chr.22:50505720 - 50603696 on Build GRCh38 | Gain |
|
| nsv834239 | Chr.22:50485317 - 50630215 on Build GRCh38 | Loss |
|
| nsv589223 | Chr.22:50539595 - 50619495 on Build GRCh38 | Loss |
|
| nsv589222 | Chr.22:50491152 - 50589773 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map