Assay Details
Target Gene Details
Entrez Gene ID: | 92552 |
Gene Name: | ataxin 3 like |
Gene Aliases: |
MJDL |
Location: |
Chr.X:13318647-13320399 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATXN3L | NM_001135995.1 | 1 | 318 | NP_001129467.1 |
| AK302365.1 | 1 | 320 | ||
| BC137186.1 | 1 | 52 | ||
| BC137187.1 | 1 | 52 | ||
| DB059638.1 | 1 | 320 | ||
| DB460385.1 | 1 | 328 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576773 | Chr.X:13071156 - 13600068 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map