Assay Details
Target Gene Details
Entrez Gene ID: | 441518 |
Gene Name: | family with sequence similarity 127 member C |
Gene Aliases: |
CXX1c, MAR8B |
Location: |
Chr.X:135020504-135022536 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM127C | NM_001078173.1 | 1 | 1541 | NP_001071641.1 |
| AK098443.1 | 1 | 1014 |
More Information
Set Membership: |
|

Genomic Map