Assay Details
Target Gene Details
Entrez Gene ID: | 101928259 |
Gene Name: | uncharacterized LOC101928259 |
Gene Aliases: |
- |
Location: |
Chr.X:72688949-72712348 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928259 | NR_110391.1 | |||
| AI968194.1 |
Target Gene Details
Entrez Gene ID: | 5255 |
Gene Name: | phosphorylase kinase regulatory subunit alpha 1 |
Gene Aliases: |
PHKA |
Location: |
Chr.X:72578814-72714181 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHKA1 | NM_001122670.1 | NP_001116142.1 | ||
| NM_001172436.1 | NP_001165907.1 | |||
| NM_002637.3 | NP_002628.2 | |||
| XM_006724661.2 | XP_006724724.1 | |||
| BC104944.1 | AAI04945.1 | |||
| BC143499.1 | ||||
| BC143501.1 | ||||
| DR002787.1 | ||||
| X73874.1 | CAA52083.1 |
More Information
Set Membership: |
|

Genomic Map