Assay Details
Target Gene Details
Entrez Gene ID: | 552891 |
Gene Name: | DNAJC25-GNG10 readthrough |
Gene Aliases: |
- |
Location: |
Chr.9:111631352-111670246 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC25-GNG10 | NM_004125.3 | NP_004116.2 | ||
| BC015391.1 | AAH15391.1 | |||
| BT007068.1 | AAP35731.1 |
Target Gene Details
Entrez Gene ID: | 2790 |
Gene Name: | G protein subunit gamma 10 |
Gene Aliases: |
- |
Location: |
Chr.9:111661571-111670246 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNG10 | NM_001017998.3 | NP_001017998.1 | ||
| NM_001198664.1 | NP_001185593.1 | |||
| AF493877.1 | AAM12591.1 | |||
| BC010384.1 | AAH10384.1 | |||
| BC016319.1 | AAH16319.1 | |||
| BC072671.1 | AAH72671.1 | |||
| BC107574.1 | AAI07575.1 | |||
| BG708408.1 | ||||
| DB504443.1 | ||||
| U31383.1 | AAC50205.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7700n100 | Chr.9:111660057 - 111716743 on Build GRCh38 | Gain |
|
| nsv825041 | Chr.9:111607177 - 111673958 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79430314] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map