Assay Details
Target Gene Details
Entrez Gene ID: | 5224 |
Gene Name: | phosphoglycerate mutase 2 |
Gene Aliases: |
GSD10, PGAM-M, PGAMM |
Location: |
Chr.7:44062727-44065587 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGAM2 | NM_000290.3 | 1 | 140 | NP_000281.2 |
| BC001904.1 | 1 | 148 | AAH01904.1 | |
| BC073741.1 | 1 | 136 | AAH73741.1 | |
| CT002473.1 | 1 | 140 | ||
| M18172.1 | 1 | 117 | AAA60072.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv8082 | Chr.7:43956156 - 44207229 on Build GRCh38 | Gain |
|
| nsv428165 | Chr.7:43855828 - 44172960 on Build GRCh38 | Gain |
|
| nsv470210 | Chr.7:44045936 - 44139229 on Build GRCh38 | Loss |
|
| nsv830980 | Chr.7:43865998 - 44078539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map