Assay Details
Target Gene Details
Entrez Gene ID: | 375612 |
Gene Name: | lipoma HMGIC fusion partner-like 3 |
Gene Aliases: |
LHFPL4 |
Location: |
Chr.7:104328656-104908558 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHFPL3 | NM_199000.2 | NP_945351.1 | ||
| XM_005250327.1 | XP_005250384.1 | |||
| AK124251.1 | ||||
| AK299759.1 | ||||
| AY260763.1 | AAP14954.1 | |||
| BC129937.1 | ||||
| BC129938.1 | ||||
| BC129941.1 |
Target Gene Details
Entrez Gene ID: | 645591 |
Gene Name: | LHFPL3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:104796506-104804092 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHFPL3-AS1 | NR_034141.1 | |||
| NR_034142.1 | 3 | 773 | ||
| BU161581.1 | 3 | 773 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5887 | Chr.7:104760803 - 104851157 on Build GRCh38 | Deletion |
|
| dgv425n21 | Chr.7:104770826 - 104846260 on Build GRCh38 | Gain |
|
| esv2660132 | Chr.7:104799604 - 104801341 on Build GRCh38 | Deletion |
|
| nsv1019625 | Chr.7:104788924 - 104821040 on Build GRCh38 | Gain |
|
| dgv6545n100 | Chr.7:104768944 - 104838640 on Build GRCh38 | Gain |
|
| nsv831086 | Chr.7:104782056 - 104956319 on Build GRCh38 | Loss |
|

Genomic Map