Assay Details
Target Gene Details
Entrez Gene ID: | 64764 |
Gene Name: | cAMP responsive element binding protein 3 like 2 |
Gene Aliases: |
BBF2H7 |
Location: |
Chr.7:137874979-138002101 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREB3L2 | NM_001253775.1 | NP_001240704.1 | ||
| NM_194071.3 | NP_919047.2 | |||
| AJ549092.1 | CAD79342.1 | |||
| AJ549387.1 | CAD79347.1 | |||
| AK131517.1 | BAD18659.1 | |||
| AK293048.1 | ||||
| BC063666.1 | AAH63666.1 | |||
| BC110813.1 | AAI10814.1 | |||
| BC131723.1 | ||||
| BQ711676.1 | ||||
| CN394705.1 | ||||
| CR749563.1 | CAH18360.1 |
Target Gene Details
Entrez Gene ID: | 100130880 |
Gene Name: | uncharacterized LOC100130880 |
Gene Aliases: |
- |
Location: |
Chr.7:137953348-137957966 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100130880 | NM_001243523.1 | NP_001230452.1 | ||
| DA296601.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831150 | Chr.7:137815813 - 137968062 on Build GRCh38 | Gain+Loss |
|
| esv33947 | Chr.7:137235266 - 138283104 on Build GRCh38 | Gain |
|
| nsv518070 | Chr.7:137849881 - 138124093 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76941256] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map