Assay Details
Target Gene Details
Entrez Gene ID: | 9715 |
Gene Name: | family with sequence similarity 131 member B |
Gene Aliases: |
- |
Location: |
Chr.7:143353400-143382304 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM131B | XM_005250073.3 | XP_005250130.1 |
Target Gene Details
Entrez Gene ID: | 100507507 |
Gene Name: | uncharacterized LOC100507507 |
Gene Aliases: |
- |
Location: |
Chr.7:143379689-143381025 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507507 | NM_001322263.1 | 1 | 417 | NP_001309192.1 |
| AI066553.1 | 1 | 75 | ||
| HY027225.1 | 1 | 417 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv608681 | Chr.7:143372217 - 143425364 on Build GRCh38 | Loss |
|
| nsv517520 | Chr.7:143352880 - 143400007 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map