Assay Details
Target Gene Details
Entrez Gene ID: | 107 |
Gene Name: | adenylate cyclase 1 |
Gene Aliases: |
AC1, DFNB44 |
Location: |
Chr.7:45574140-45723116 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY1 | NM_001281768.1 | NP_001268697.1 | ||
| NM_021116.2 | 1 | 195 | NP_066939.1 | |
| XM_005249584.3 | 1 | 193 | XP_005249641.1 | |
| XM_005249585.2 | 1 | 193 | XP_005249642.1 | |
| AB209496.1 | 1 | 140 | BAD92733.1 | |
| AF497515.1 | 1 | 199 | AAM94372.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1076890 | Chr.7:45574300 - 45574701 on Build GRCh38 | Deletion |
|
| esv24848 | Chr.7:45574204 - 45574919 on Build GRCh38 | Loss |
|
| nsv950210 | Chr.7:45572902 - 45578501 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map