Assay Details
Target Gene Details
Entrez Gene ID: | 51809 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 7 |
Gene Aliases: |
GALNAC-T7, GalNAcT7 |
Location: |
Chr.4:173168753-173323967 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT7 | NM_017423.2 | 1 | 158 | NP_059119.2 |
| XM_005263062.1 | 1 | 158 | XP_005263119.1 | |
| AJ002744.1 | 1 | 107 | CAB60270.1 | |
| BC046129.1 | 1 | 136 | AAH46129.1 | |
| BC047468.1 | 1 | 133 | AAH47468.1 | |
| DB238519.1 | 1 | 158 |
Target Gene Details
Entrez Gene ID: | 101930370 |
Gene Name: | uncharacterized LOC101930370 |
Gene Aliases: |
- |
Location: |
Chr.4:173131933-173169652 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101930370 | NR_134241.1 | 1 | 743 | |
| NR_134242.1 | 1 | 743 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv24067 | Chr.4:173168863 - 173170026 on Build GRCh38 | Loss |
|
| nsv1074239 | Chr.4:173168648 - 173169249 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map