Assay Details
Target Gene Details
Entrez Gene ID: | 64816 |
Gene Name: | cytochrome P450 family 3 subfamily A member 43 |
Gene Aliases: |
- |
Location: |
Chr.7:99828013-99867138 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP3A43 | NM_001278921.1 | NP_001265850.1 | ||
| NM_022820.4 | NP_073731.1 | |||
| NM_057095.2 | NP_476436.1 | |||
| NM_057096.3 | NP_476437.1 | |||
| NR_103868.1 | ||||
| NR_103869.1 | ||||
| XM_017012542.1 | XP_016868031.1 | |||
| XM_017012543.1 | XP_016868032.1 | |||
| XM_017012544.1 | XP_016868033.1 | |||
| XM_017012545.1 | XP_016868034.1 | |||
| AA417369.1 | ||||
| AF280108.1 | AAG33009.1 | |||
| AF280109.1 | AAG33010.1 | |||
| AF280110.1 | AAG33011.1 | |||
| AF280111.1 | AAG33012.1 | |||
| AF280112.1 | AAG33013.1 | |||
| AF280113.1 | AAG33014.1 | |||
| AF319634.1 | AAK00325.1 | |||
| AF337813.1 | AAK38841.1 | |||
| AY390423.1 | AAQ92351.1 | |||
| AY390424.1 | AAQ92352.1 | |||
| AY390425.1 | AAQ92353.1 | |||
| AY390426.1 | AAQ92354.1 | |||
| BC100981.3 | AAI00982.1 | |||
| BC100982.3 | AAI00983.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3614285 | Chr.7:99863718 - 99865907 on Build GRCh38 | Loss |
|
| esv29538 | Chr.7:99863828 - 99865924 on Build GRCh38 | Loss |
|
| esv3542152 | Chr.7:99863758 - 99865939 on Build GRCh38 | Deletion |
|
| esv2670223 | Chr.7:99863766 - 99865939 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map