Assay Details
Target Gene Details
Entrez Gene ID: | 340512 |
Gene Name: | uncharacterized LOC340512 |
Gene Aliases: |
- |
Location: |
Chr.9:106974833-107102988 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC340512 | NR_126029.1 | |||
| AK097706.1 |
Target Gene Details
Entrez Gene ID: | 58499 |
Gene Name: | zinc finger protein 462 |
Gene Aliases: |
Zfp462 |
Location: |
Chr.9:106860154-107012258 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF462 | NM_021224.4 | NP_067047.4 | ||
| XM_006717209.3 | XP_006717272.1 | |||
| XM_006717211.3 | XP_006717274.1 | |||
| XM_006717212.3 | XP_006717275.1 | |||
| XM_006717215.3 | XP_006717278.1 | |||
| XM_006717216.3 | XP_006717279.1 | |||
| XM_006717218.3 | XP_006717281.1 | |||
| XM_011518892.2 | XP_011517194.1 | |||
| XM_017014996.1 | XP_016870485.1 | |||
| XM_017014997.1 | XP_016870486.1 | |||
| XM_017014998.1 | XP_016870487.1 | |||
| XM_017014999.1 | XP_016870488.1 | |||
| AB058706.2 | BAB47432.2 | |||
| AK027866.1 | ||||
| AK302007.1 | ||||
| BC036884.1 | AAH36884.1 | |||
| BX648965.1 | CAH56168.1 | |||
| CR749850.1 | CAH18698.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621392 | Chr.9:106977210 - 106979773 on Build GRCh38 | Loss |
|
| nsv831680 | Chr.9:106891315 - 107086546 on Build GRCh38 | Loss |
|
| esv2673055 | Chr.9:106977212 - 106979771 on Build GRCh38 | Deletion |
|
| esv29808 | Chr.9:106977301 - 106979792 on Build GRCh38 | Loss |
|
| nsv968743 | Chr.9:106977369 - 106979277 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map