Assay Details
Target Gene Details
Entrez Gene ID: | 23232 |
Gene Name: | TBC1 domain family member 12 |
Gene Aliases: |
- |
Location: |
Chr.10:94402429-94536332 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D12 | NM_015188.1 | 1 | 528 | NP_056003.1 |
| XM_006717732.3 | 1 | 528 | XP_006717795.2 | |
| XM_017015996.1 | 1 | 528 | XP_016871485.1 | |
| XM_017015997.1 | 1 | 528 | XP_016871486.1 | |
| AB011180.1 | 1 | 362 | ||
| AB449898.1 | 1 | 343 | ||
| DA808973.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv25061 | Chr.10:94402654 - 94403589 on Build GRCh38 | Loss |
|
| nsv831946 | Chr.10:94295433 - 94466681 on Build GRCh38 | Loss |
|
| nsv551950 | Chr.10:94402684 - 94403387 on Build GRCh38 | Gain |
|
| nsv551947 | Chr.10:94394510 - 94403212 on Build GRCh38 | Loss |
|
| esv2761616 | Chr.10:94207011 - 94543944 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map