Assay Details
Target Gene Details
Entrez Gene ID: | 4666 |
Gene Name: | nascent polypeptide-associated complex alpha subunit |
Gene Aliases: |
HSD48, NAC-alpha, NACA1, skNAC |
Location: |
Chr.12:56712427-56726275 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NACA | NM_001113201.2 | NP_001106672.1 | ||
| NM_001113202.1 | NP_001106673.1 | |||
| NM_001113203.2 | 3 | 1281 | NP_001106674.2 | |
| NM_001320193.1 | NP_001307122.1 | |||
| NM_001320194.1 | NP_001307123.1 | |||
| NM_005594.5 | NP_005585.1 | |||
| XM_006719412.1 | 3 | 2040 | XP_006719475.1 | |
| XM_006719413.2 | 3 | 1296 | XP_006719476.1 | |
| XM_006719414.1 | 3 | 1038 | XP_006719477.1 | |
| XM_006719418.1 | XP_006719481.1 | |||
| XM_006719420.3 | XP_006719483.1 | |||
| XM_011538389.1 | 3 | 1038 | XP_011536691.1 | |
| XM_017019337.1 | 3 | 1038 | XP_016874826.1 | |
| AF054187.1 | AAC99403.1 | |||
| AK090650.1 | ||||
| AK096699.1 | 3 | 1038 | ||
| AK223085.1 | BAD96805.1 | |||
| AK293209.1 | ||||
| AK311904.1 | ||||
| AY034001.1 | AAK57544.1 | |||
| AY605660.1 | AAV83778.1 | |||
| AY911673.1 | AAX14393.1 | |||
| BC105120.1 | AAI05121.1 | |||
| BC105122.1 | AAI05123.1 | |||
| BC106041.1 | AAI06042.1 | |||
| BG709003.1 | ||||
| BI856347.1 | ||||
| CD104788.1 | ||||
| CD246364.1 | ||||
| CR450295.1 | CAG29291.1 | |||
| DB487006.1 | ||||
| X80909.1 | CAA56869.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
| esv22791 | Chr.12:56716780 - 56721722 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map