Assay Details
Target Gene Details
Entrez Gene ID: | 100287082 |
Gene Name: | gastric cancer associated transcript 2 (non-protein coding) |
Gene Aliases: |
MTCL1-AS1, MTCL1AS1 |
Location: |
Chr.18:8695856-8707621 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GACAT2 | NR_120598.1 | |||
| BM717007.1 | ||||
| BQ026945.1 |
Target Gene Details
Entrez Gene ID: | 23255 |
Gene Name: | microtubule crosslinking factor 1 |
Gene Aliases: |
CCDC165, KIAA0802, SOGA2 |
Location: |
Chr.18:8705576-8832777 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTCL1 | XM_005258098.3 | 1 | 701 | XP_005258155.1 |
| XM_005258099.4 | 1 | 701 | XP_005258156.1 | |
| XM_011525640.2 | 1 | 701 | XP_011523942.1 | |
| XM_011525641.2 | 1 | 701 | XP_011523943.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23442 | Chr.18:8704955 - 8706815 on Build GRCh38 | Loss |
|
| dgv5843n54 | Chr.18:8701637 - 8706759 on Build GRCh38 | Loss |
|
| dgv5844n54 | Chr.18:8705082 - 8706635 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map