Assay Details
Target Gene Details
Entrez Gene ID: | 388552 |
Gene Name: | biogenesis of lysosomal organelles complex 1 subunit 3 |
Gene Aliases: |
BLOS3, HPS8, RP |
Location: |
Chr.19:45178745-45202715 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLOC1S3 | NM_212550.4 | NP_997715.1 | ||
| AK296695.1 | ||||
| DC422050.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422482 | Chr.19:45173447 - 45266547 on Build GRCh38 | Duplication |
|
| esv29928 | Chr.19:45178250 - 45178760 on Build GRCh38 | Loss |
|
| esv1005203 | Chr.19:45172554 - 45181610 on Build GRCh38 | Deletion |
|
| nsv833843 | Chr.19:45090630 - 45290424 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map