Assay Details
Target Gene Details
Entrez Gene ID: | 79090 |
Gene Name: | trafficking protein particle complex 6A |
Gene Aliases: |
TRS33 |
Location: |
Chr.19:45162928-45178302 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAPPC6A | XM_017027295.1 | XP_016882784.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422482 | Chr.19:45173447 - 45266547 on Build GRCh38 | Duplication |
|
| esv29928 | Chr.19:45178250 - 45178760 on Build GRCh38 | Loss |
|
| esv1005203 | Chr.19:45172554 - 45181610 on Build GRCh38 | Deletion |
|
| nsv833843 | Chr.19:45090630 - 45290424 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map