Assay Details
Target Gene Details
Entrez Gene ID: | 23141 |
Gene Name: | ankyrin repeat and LEM domain containing 2 |
Gene Aliases: |
KIAA0692, LEMD7, Lem4, MCPH16 |
Location: |
Chr.12:132725667-132761888 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKLE2 | NM_015114.2 | 11 | 2399 | NP_055929.1 |
| XM_005266159.3 | 11 | 2320 | XP_005266216.1 | |
| XM_005266160.1 | 11 | 2256 | XP_005266217.1 | |
| XM_006719735.1 | XP_006719798.1 | |||
| XM_017019076.1 | 12 | 2339 | XP_016874565.1 | |
| AB014592.1 | 10 | 1846 | BAA31667.1 | |
| AK024061.1 | 6 | 1087 | ||
| AK025933.1 | 1 | 94 | ||
| AK093451.1 | 2 | 1833 | ||
| AK293028.1 | 2 | 723 | ||
| BC043157.1 | 11 | 2345 | AAH43157.2 | |
| BC062373.1 | 1 | 407 | AAH62373.1 | |
| BC082962.1 | 1 | 407 | AAH82962.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580552 | Chr.12:132728124 - 132732931 on Build GRCh38 | Loss |
|
| nsv561128 | Chr.12:132729570 - 132730284 on Build GRCh38 | Loss |
|
| nsv561122 | Chr.12:132726449 - 132729875 on Build GRCh38 | Loss |
|
| nsv1047500 | Chr.12:132666737 - 132763400 on Build GRCh38 | Gain |
|
| nsv1037074 | Chr.12:132613063 - 132801953 on Build GRCh38 | Gain |
|
| nsv561125 | Chr.12:132729415 - 132730003 on Build GRCh38 | Gain+Loss |
|
| esv21605 | Chr.12:132729386 - 132730388 on Build GRCh38 | Gain |
|
| esv3631311 | Chr.12:132713141 - 132737944 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map