Assay Details
Target Gene Details
Entrez Gene ID: | 10389 |
Gene Name: | sex comb on midleg-like 2 (Drosophila) |
Gene Aliases: |
- |
Location: |
Chr.X:18239313-18355089 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCML2 | NM_006089.2 | NP_006080.1 | ||
| NR_033717.1 | ||||
| XM_006724458.1 | XP_006724521.1 | |||
| XM_017029219.1 | XP_016884708.1 | |||
| XM_017029220.1 | XP_016884709.1 | |||
| XM_017029221.1 | XP_016884710.1 | |||
| XM_017029222.1 | XP_016884711.1 | |||
| AK299190.1 | ||||
| AK303064.1 | ||||
| AK308140.1 | ||||
| AK315116.1 | ||||
| BC051913.1 | AAH51913.1 | |||
| BC064617.1 | AAH64617.1 | |||
| Y18004.1 | CAB38943.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2663102 | Chr.X:18260644 - 18262818 on Build GRCh38 | Deletion |
|
| esv24903 | Chr.X:18259540 - 18263301 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map