Assay Details
Target Gene Details
Entrez Gene ID: | 124930 |
Gene Name: | ankyrin repeat domain 13B |
Gene Aliases: |
- |
Location: |
Chr.17:29589769-29614761 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD13B | XM_017024174.1 | XP_016879663.1 | ||
| BC062597.1 | AAH62597.1 | |||
| DA445860.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521557 | Chr.17:29562968 - 29595257 on Build GRCh38 | Loss |
|
| nsv833409 | Chr.17:29465646 - 29655807 on Build GRCh38 | Gain |
|
| esv25114 | Chr.17:29589029 - 29589913 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111418919] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map