Assay Details
Target Gene Details
Entrez Gene ID: | 728763 |
Gene Name: | ciliary rootlet coiled-coil, rootletin family member 2 |
Gene Aliases: |
- |
Location: |
Chr.2:240919048-240993311 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CROCC2 | XM_017005450.1 | XP_016860939.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584861 | Chr.2:240810727 - 241018260 on Build GRCh38 | Gain |
|
| esv29869 | Chr.2:240923301 - 240925519 on Build GRCh38 | Gain+Loss |
|
| nsv584935 | Chr.2:240924422 - 240933180 on Build GRCh38 | Loss |
|
| nsv511162 | Chr.2:240921494 - 240933305 on Build GRCh38 | Loss |
|
| dgv7382n54 | Chr.2:240923534 - 240933305 on Build GRCh38 | Gain |
|
| nsv470531 | Chr.2:240684476 - 240928657 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map