Assay Details
Target Gene Details
Entrez Gene ID: | 728763 |
Gene Name: | ciliary rootlet coiled-coil, rootletin family member 2 |
Gene Aliases: |
- |
Location: |
Chr.2:240919048-240993311 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CROCC2 | XM_017005450.1 | XP_016860939.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584861 | Chr.2:240810727 - 241018260 on Build GRCh38 | Gain |
|
| nsv584940 | Chr.2:240971610 - 241040706 on Build GRCh38 | Loss |
|
| nsv820947 | Chr.2:240975190 - 240983719 on Build GRCh38 | Deletion |
|
| esv3363419 | Chr.2:240978762 - 240981360 on Build GRCh38 | Duplication |
|
| esv22673 | Chr.2:240975448 - 240982779 on Build GRCh38 | Gain+Loss |
|
| nsv821941 | Chr.2:240969592 - 240982457 on Build GRCh38 | Loss |
|
| dgv7388n54 | Chr.2:240976067 - 240981327 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map