Assay Details
Target Gene Details
Entrez Gene ID: | 51809 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 7 |
Gene Aliases: |
GALNAC-T7, GalNAcT7 |
Location: |
Chr.4:173168753-173323967 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT7 | NM_017423.2 | NP_059119.2 | ||
| XM_005263062.1 | XP_005263119.1 | |||
| AJ002744.1 | CAB60270.1 | |||
| BC046129.1 | AAH46129.1 | |||
| BC047468.1 | AAH47468.1 | |||
| DB238519.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv24067 | Chr.4:173168863 - 173170026 on Build GRCh38 | Loss |
|
| nsv596232 | Chr.4:173169375 - 173169959 on Build GRCh38 | Gain |
|
| dgv9397n54 | Chr.4:173169007 - 173169969 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map