Assay Details
Target Gene Details
Entrez Gene ID: | 381091 |
Gene Name: | histocompatibility 2, class II antigen E beta2 |
Gene Aliases: |
A130038H09Rik, H-2Eb2, Ia-5, Ia5, Iebeta2 |
Location: |
Chr.17:34325500-34341410 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2-Eb2 | NM_001033978.3 | 2 | 212 | NP_001029150.1 |
| AK137912.1 | 2 | 213 | BAE23514.1 | |
| AK157199.1 | 2 | 223 | BAE33995.1 | |
| BC150834.1 | 2 | 213 | ||
| KJ650234.1 | 2 | 195 |

Genomic Map