Assay Details
Target Gene Details
Entrez Gene ID: | 12408 |
Gene Name: | carbonyl reductase 1 |
Gene Aliases: |
AW261796, CR, Cbr |
Location: |
Chr.16:93607837-93610349 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cbr1 | NM_007620.2 | 1 | 78 | NP_031646.2 |
| AK146805.1 | ||||
| BC012714.1 | AAH12714.1 | |||
| BC158026.1 | 1 | 56 | ||
| BC158029.1 | 1 | 56 | ||
| BY319589.1 | 1 | 79 | ||
| U31966.1 | 1 | 77 | AAB19006.1 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map