Assay Details
Target Gene Details
Entrez Gene ID: | 14961 |
Gene Name: | histocompatibility 2, class II antigen A, beta 1 |
Gene Aliases: |
AI845868, Abeta, H-2Ab, H2-Ab, I-Abeta, IAb, Ia-2, Ia2, Rmcs1 |
Location: |
Chr.17:34263227-34269418 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2-Ab1 | NM_207105.3 | 6 | 1039 | NP_996988.2 |
| AK077436.1 | 6 | 1027 | BAC36800.1 | |
| AK155573.1 | 6 | 1011 | BAE33331.1 | |
| AK156899.1 | 6 | 1011 | BAE33890.1 | |
| AK170370.1 | 6 | 1021 | BAE41751.1 | |
| AK171005.1 | 6 | 1011 | BAE42176.1 | |
| AK171211.1 | 6 | 1015 | BAE42316.1 | |
| AK171281.1 | 6 | 1011 | BAE42364.1 | |
| AK171290.1 | 6 | 1011 | BAE42372.1 | |
| AK171298.1 | 6 | 1011 | BAE42377.1 | |
| AK172409.1 | 6 | 1012 | ||
| BB056841.1 | 1 | 86 | ||
| BC008168.1 | 6 | 1031 | AAH08168.1 | |
| BC010322.1 | 6 | 1005 | AAH10322.1 | |
| BC057998.1 | 6 | 997 | AAH57998.1 | |
| CK330363.1 | 1 | 147 | ||
| K01143.1 | 3 | 314 | AAA39545.1 |
More Information
Set Membership: |
|

Genomic Map