Assay Details
Target Gene Details
Entrez Gene ID: | 15980 |
Gene Name: | interferon gamma receptor 2 |
Gene Aliases: |
Ifgr2, Ifgt |
Location: |
Chr.16:91547094-91564007 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ifngr2 | NM_008338.3 | NP_032364.1 | ||
| AK086929.1 | BAC39764.1 | |||
| AK137165.1 | BAE23260.1 | |||
| AK143214.1 | BAE25310.1 | |||
| AK165193.1 | BAE38071.1 | |||
| AK169907.1 | ||||
| BC055745.1 | AAH55745.1 | |||
| S69336.1 | AAB30165.1 |
More Information
Set Membership: |
|

Genomic Map