Assay Details
Target Gene Details
Entrez Gene ID: | 26572 |
Gene Name: | COP9 (constitutive photomorphogenic) homolog, subunit 3 (Arabidopsis thaliana) |
Gene Aliases: |
Csn3, Sgn3 |
Location: |
Chr.11:59817795-59839844 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cops3 | NM_011991.1 | NP_036121.1 | ||
| AF071313.1 | AAC33900.1 | |||
| AK142318.1 | BAE25027.1 | |||
| AK169747.1 | BAE41343.1 | |||
| BC068179.1 | AAH68179.1 |
Target Gene Details
Entrez Gene ID: | 100504104 |
Gene Name: | predicted gene 16062 |
Gene Aliases: |
ENSMUSG00000072890, OTTMUSG00000029633 |
Location: |
Chr.11:59810080-59818362 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm16062 | NR_045686.1 | |||
| AK144909.1 | BAE26130.1 |
More Information
Set Membership: |
|

Genomic Map