Assay Details
Target Gene Details
Entrez Gene ID: | 26572 |
Gene Name: | COP9 (constitutive photomorphogenic) homolog, subunit 3 (Arabidopsis thaliana) |
Gene Aliases: |
Csn3, Sgn3 |
Location: |
Chr.11:59817795-59839844 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cops3 | NM_011991.1 | 2 | 159 | NP_036121.1 |
| AF071313.1 | 2 | 159 | AAC33900.1 | |
| AK048906.1 | 2 | 149 | BAC33487.1 | |
| AK142318.1 | 2 | 145 | BAE25027.1 | |
| BC068179.1 | 2 | 159 | AAH68179.1 |
More Information
Set Membership: |
|

Genomic Map