Assay Details
Target Gene Details
Entrez Gene ID: | 14969 |
Gene Name: | histocompatibility 2, class II antigen E beta |
Gene Aliases: |
H-2Eb, H2Eb, Ia-4, Ia4 |
Location: |
Chr.17:34305867-34316674 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2-Eb1 | NM_010382.2 | NP_034512.2 | ||
| AK155968.1 | BAE33527.1 | |||
| AK160485.1 | BAE35816.1 | |||
| BC132163.1 | ||||
| BC132165.1 | ||||
| K01145.1 | AAA39594.1 | |||
| M11355.1 | AAA39644.1 | |||
| M12826.1 | AAA39645.1 | |||
| M28591.1 | AAA39589.1 | |||
| M36939.1 | AAA39593.1 | |||
| M36940.1 | AAA39590.1 | |||
| U28489.1 | AAB17688.1 | |||
| U28490.1 | AAB17689.1 | |||
| X00958.1 | CAA25470.1 | |||
| X52641.1 | CAA36863.1 | |||
| X52642.1 | CAA36864.1 |
More Information
Set Membership: |
|

Genomic Map