Assay Details
Target Gene Details
Entrez Gene ID: | 109857 |
Gene Name: | carbonyl reductase 3 |
Gene Aliases: |
1110001J05Rik, C81353 |
Location: |
Chr.16:93683219-93690991 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cbr3 | NM_173047.3 | NP_766635.1 | ||
| AK003232.1 | ||||
| AK028150.1 | BAC25778.1 | |||
| BC028763.1 | AAH28763.1 | |||
| BC087735.1 | AAH87735.1 | |||
| BC096658.1 | AAH96658.1 |
More Information
Set Membership: |
|

Genomic Map