Assay Details
Target Gene Details
Entrez Gene ID: | 14961 |
Gene Name: | histocompatibility 2, class II antigen A, beta 1 |
Gene Aliases: |
AI845868, Abeta, H-2Ab, H2-Ab, I-Abeta, IAb, Ia-2, Ia2, Rmcs1 |
Location: |
Chr.17:34263227-34269418 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2-Ab1 | NM_207105.3 | 1 | 130 | NP_996988.2 |
| AF065912.1 | AAC17910.1 | |||
| AF065913.1 | 1 | 66 | AAC17911.1 | |
| AF119251.1 | AAD31754.1 | |||
| AF119252.1 | AAD31755.1 | |||
| AK077436.1 | 1 | 118 | BAC36800.1 | |
| AK155573.1 | 1 | 108 | BAE33331.1 | |
| AK156899.1 | 1 | 108 | BAE33890.1 | |
| AK170370.1 | 1 | 112 | BAE41751.1 | |
| AK171005.1 | 1 | 108 | BAE42176.1 | |
| AK171211.1 | 1 | 112 | BAE42316.1 | |
| AK171281.1 | 1 | 108 | BAE42364.1 | |
| AK171290.1 | 1 | 108 | BAE42372.1 | |
| AK171298.1 | 1 | 108 | BAE42377.1 | |
| AK172409.1 | 1 | 108 | ||
| AY452202.1 | AAR19090.1 | |||
| BC008168.1 | 1 | 128 | AAH08168.1 | |
| BC010322.1 | 1 | 96 | AAH10322.1 | |
| BC057998.1 | 1 | 94 | AAH57998.1 | |
| BY205244.1 | 1 | 149 | ||
| M13537.1 | AAA39633.1 | |||
| M13538.1 | AAA39632.1 | |||
| M13539.1 | AAA39635.1 | |||
| M13540.1 | AAA39634.1 | |||
| M13541.1 | AAA39629.1 | |||
| M15848.1 | 1 | 81 | AAA39547.1 | |
| M66213.1 | AAA20652.1 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map