Assay Details
Target Gene Details
Entrez Gene ID: | 114604 |
Gene Name: | PR domain containing 15 |
Gene Aliases: |
C21orf83, E130018M06Rik, ORF62, Zfp298 |
Location: |
Chr.16:97791467-97852427 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prdm15 | NM_144789.2 | NP_659038.2 | ||
| AK144820.1 | BAE26081.1 |
More Information
Set Membership: |
|

Genomic Map