Assay Details
Target Gene Details
Entrez Gene ID: | 114604 |
Gene Name: | PR domain containing 15 |
Gene Aliases: |
C21orf83, E130018M06Rik, ORF62, Zfp298 |
Location: |
Chr.16:97791467-97852427 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prdm15 | NM_144789.2 | 23 | 3049 | NP_659038.2 |
| AK053449.1 | 1 | 104 | ||
| AK144820.1 | 23 | 2954 | BAE26081.1 | |
| AY063457.1 | 4 | 607 | AAL60597.1 | |
| BC046433.1 | 4 | 608 | AAH46433.1 |
More Information
Set Membership: |
|

Genomic Map