Assay Details
Target Gene Details
Entrez Gene ID: | 23872 |
Gene Name: | E26 avian leukemia oncogene 2, 3' domain |
Gene Aliases: |
AU022856, Ets-2 |
Location: |
Chr.16:95702407-95721049 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ets2 | NM_011809.3 | 8 | 1179 | NP_035939.3 |
| AK014602.2 | 8 | 1101 | ||
| AK030537.1 | 4 | 481 | BAC27010.1 | |
| AK038348.1 | 8 | 1166 | BAE20535.1 | |
| AK128933.1 | 9 | 1053 | ||
| AK143687.1 | 8 | 1131 | BAE25498.1 | |
| AK144974.1 | 8 | 1130 | BAE26161.1 | |
| AK145852.1 | 8 | 1015 | BAE26698.1 | |
| AK150175.1 | 8 | 1108 | ||
| AK151315.1 | 8 | 1130 | BAE30296.1 | |
| AK152750.1 | 8 | 1133 | ||
| AK153339.1 | 8 | 1131 | BAE31917.1 | |
| AK153478.1 | 8 | 1131 | BAE32028.1 | |
| AK183948.1 | ||||
| BC005486.1 | 8 | 1100 | AAH05486.1 | |
| BC005504.1 | 8 | 1040 | AAH05504.1 | |
| CT010235.1 | 7 | 941 | CAJ18443.1 | |
| J04103.1 | 8 | 1078 | AAA37581.1 |
More Information
Set Membership: |
|

Genomic Map