Assay Details
Target Gene Details
Entrez Gene ID: | 13495 |
Gene Name: | developmentally regulated GTP binding protein 2 |
Gene Aliases: |
AI255295, AI461605 |
Location: |
Chr.11:60454617-60468724 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Drg2 | NM_021354.1 | NP_067329.1 | ||
| AJ243590.1 | CAB65258.1 | |||
| AK012988.1 | ||||
| AK197050.1 | ||||
| BC082564.1 | AAH82564.1 |

Genomic Map