Assay Details
Target Gene Details
Entrez Gene ID: | 12314 |
Gene Name: | calmodulin 2 |
Gene Aliases: |
1500001E21Rik, AL024017 |
Location: |
Chr.17:87433401-87446935 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Calm2 | NM_007589.5 | NP_031615.1 | ||
| AK012247.1 | BAB28116.1 | |||
| AK012564.1 | BAB28319.1 | |||
| AK013068.1 | BAB28631.1 | |||
| AK152303.1 | BAE31109.1 | |||
| AK160636.1 | BAE35930.1 | |||
| AK161268.1 | BAE36280.1 | |||
| AK161984.1 | BAE36667.1 | |||
| AK168741.1 | BAE40582.1 | |||
| BC010730.1 | AAH10730.1 | |||
| BC021347.1 | AAH21347.1 | |||
| BC051444.1 | AAH51444.1 | |||
| BC100301.1 | AAI00302.1 | |||
| L31642.1 | AAA65934.1 |
More Information
Set Membership: |
|

Genomic Map