Assay Details
Target Gene Details
Entrez Gene ID: | 12929 |
Gene Name: | v-crk sarcoma virus CT10 oncogene homolog (avian)-like |
Gene Aliases: |
1110025F07Rik, AA589403, AI325100, Crkol |
Location: |
Chr.16:17451985-17487440 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crkl | NM_001277231.1 | 1 | 165 | NP_001264160.1 |
| NM_007764.5 | 1 | 165 | NP_031790.2 | |
| AK048641.1 | 1 | 163 | BAC33406.1 | |
| AK052315.1 | 1 | 165 | BAC34933.1 | |
| AK163981.1 | 1 | 137 | BAE37567.1 | |
| X90648.1 | CAA62220.1 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map