Assay Details
Target Gene Details
Entrez Gene ID: | 13555 |
Gene Name: | E2F transcription factor 1 |
Gene Aliases: |
E2F-1, mKIAA4009 |
Location: |
Chr.2:154559400-154569892 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E2f1 | NM_001291105.1 | NP_001278034.1 | ||
| NM_007891.5 | NP_031917.1 | |||
| AF483514.1 | AAL90788.1 | |||
| AF483515.1 | AAL90789.1 | |||
| AK017841.1 | BAB30968.1 | |||
| AK153968.1 | BAE32285.1 | |||
| AK182005.1 | ||||
| AK211069.1 | ||||
| AK220226.1 | BAD90151.1 | |||
| BC052160.1 | AAH52160.2 | |||
| BY254403.1 | ||||
| L21973.1 | AAA83217.1 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map