Assay Details
Target Gene Details
Entrez Gene ID: | 68736 |
Gene Name: | tRNA-yW synthesizing protein 5 |
Gene Aliases: |
1110034B05Rik |
Location: |
Chr.1:57387899-57406674 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tyw5 | NM_001037742.2 | NP_001032831.2 | ||
| AK139666.1 | BAE24098.1 | |||
| BC132289.1 | ||||
| BU697704.1 | ||||
| CF951173.1 |

Genomic Map