Assay Details
Target Gene Details
Entrez Gene ID: | 15463 |
Gene Name: | ArfGAP with FG repeats 1 |
Gene Aliases: |
AU045498, C130049H11Rik, C85612, D730048C23Rik, Hrb, RAB, Rip |
Location: |
Chr.1:82839460-82901001 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agfg1 | NM_010472.2 | NP_034602.1 | ||
| AF057287.1 | AAC97477.1 | |||
| AK029917.1 | BAC26675.1 | |||
| AK048326.1 | BAC33303.1 | |||
| AK049400.1 | BAC33736.1 | |||
| AK198918.1 | ||||
| BC031154.1 | AAH31154.1 |
More Information
Set Membership: |
|

Genomic Map