Assay Details
Target Gene Details
Entrez Gene ID: | 11611 |
Gene Name: | alanine-glyoxylate aminotransferase |
Gene Aliases: |
AGT, Agt1, Agxt1, SPT |
Location: |
Chr.1:93135240-93145421 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agxt | NM_001276710.1 | NP_001263639.1 | ||
| NM_016702.3 | NP_057911.2 | |||
| AF027730.2 | AAB82001.2 | |||
| AK149432.1 | BAE28872.1 | |||
| BC025799.1 | AAH25799.1 |
More Information
Set Membership: |
|

Genomic Map