Assay Details
Target Gene Details
Entrez Gene ID: | 11807 |
Gene Name: | apolipoprotein A-II |
Gene Aliases: |
Alp-2, Apo-AII, ApoA-II, ApoAII, Apoa-2, Hdl-1 |
Location: |
Chr.1:171225011-171226379 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apoa2 | NM_013474.2 | NP_038502.2 | ||
| AA060885.1 | ||||
| AK145823.1 | BAE26675.1 | |||
| BY780589.1 | ||||
| CB947382.1 | ||||
| M79361.1 | AAA37249.1 | |||
| M79362.1 | AAA37250.1 | |||
| X62772.1 | CAA44616.1 |
More Information
Set Membership: |
|

Genomic Map