Assay Details
Target Gene Details
Entrez Gene ID: | 70861 |
Gene Name: | aldo-keto reductase family 1, member C-like |
Gene Aliases: |
4921521F21Rik, AI451536, AI503553 |
Location: |
Chr.1:65013078-65032770 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1cl | NM_027582.3 | NP_081858.2 | ||
| AK014937.1 | BAB29627.1 | |||
| AK135492.1 | BAE22552.1 | |||
| BC051128.1 | AAH51128.1 |
More Information
Set Membership: |
|

Genomic Map