Assay Details
Target Gene Details
Entrez Gene ID: | 12021 |
Gene Name: | BRCA1 associated RING domain 1 |
Gene Aliases: |
ENSMUSG00000060893, ENSMUSG00000073653 |
Location: |
Chr.1:71027531-71103233 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bard1 | NM_007525.3 | NP_031551.1 | ||
| AF057157.1 | AAC18095.1 | |||
| AK145528.1 | BAE26486.1 | |||
| BC128370.1 | ||||
| BC128371.1 |

Genomic Map