Assay Details
Target Gene Details
Entrez Gene ID: | 12372 |
Gene Name: | calsequestrin 1 |
Gene Aliases: |
CSQ, CSQ-1, CSQ1, sCSQ |
Location: |
Chr.1:172209894-172219895 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Casq1 | NM_009813.2 | NP_033943.2 | ||
| AK049865.1 | BAC33963.1 | |||
| BC064057.1 | ||||
| U93291.1 | AAC63616.1 |
More Information
Set Membership: |
|

Genomic Map