Assay Details
Target Gene Details
Entrez Gene ID: | 23827 |
Gene Name: | bisphosphate 3'-nucleotidase 1 |
Gene Aliases: |
BPntase |
Location: |
Chr.1:185331965-185357769 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bpnt1 | NM_011794.3 | NP_035924.2 | ||
| AF125043.1 | AAD17330.1 | |||
| AK008091.1 | ||||
| AK008714.1 | BAB25850.1 | |||
| AK093938.1 | ||||
| AK150877.1 | BAE29926.1 | |||
| AK151931.1 | BAE30807.1 | |||
| AK152009.1 | BAE30872.1 | |||
| AK154435.1 | BAE32585.1 | |||
| BC011036.1 | AAH11036.1 | |||
| BY744642.1 |
More Information
Set Membership: |
|

Genomic Map