Assay Details
Target Gene Details
Entrez Gene ID: | 70829 |
Gene Name: | coiled-coil domain containing 93 |
Gene Aliases: |
4633402D15Rik, 9230102M16Rik |
Location: |
Chr.1:121431067-121506460 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc93 | NM_001025156.2 | NP_001020327.1 | ||
| NM_029955.3 | NP_084231.2 | |||
| AK014621.1 | BAB29467.1 | |||
| AK020306.1 | BAB32065.1 | |||
| AK052300.1 | BAC34926.1 | |||
| AK082581.1 | BAC38538.1 | |||
| AK085701.1 | BAC39512.1 | |||
| AK142302.1 | BAE25021.1 | |||
| AK159415.1 | BAE35063.1 | |||
| BC054083.1 | AAH54083.1 | |||
| BY136404.1 | ||||
| BY730410.1 |

Genomic Map