Assay Details
Target Gene Details
Entrez Gene ID: | 12283 |
Gene Name: | calcium binding protein 39 |
Gene Aliases: |
AA408805, AA960512, C78372, MO25, MO25alpha |
Location: |
Chr.1:85793410-85851577 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cab39 | NM_133781.4 | NP_598542.3 | ||
| AK144665.1 | BAE25997.1 | |||
| AK187903.1 | ||||
| AK199827.1 | ||||
| BC020041.1 | AAH20041.1 | |||
| BC029053.1 | AAH29053.1 | |||
| S51858.1 | AAB24801.1 |

Genomic Map